missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol hemifumarate, MedChemExpress

Product Code. p-200099183
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16491996

Brand: Medchem Express HYB0076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bisoprolol hemifumarate is a potent, selective and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. Bisoprolol hemifumarate has the potential for hypertension, coronary artery disease and stable ventricular dysfunction research.

Bisoprolol hemifumarate is a potent, selective and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. Bisoprolol hemifumarate has the potential for hypertension, coronary artery disease and stable ventricular dysfunction research.

Specifications

Chemical Name or Material Bisoprolol hemifumarate
CAS 104344-23-2
Color White
Purity Grade Notes Research
Molecular Formula C18H31NO4.1/2C4H4O4
Quantity 500 mg
Solubility Information DMSO : ≥ 50 mg/mL (65.19 mM) ∣H2O : 20 mg/mL (26.08 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=C(COCCOC(C)C)C=C1.OC(/C=C/C(O)=O)=O.[1/2]
Molecular Weight (g/mol) 766.96
Formula Weight 766.96
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.