missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol hemifumarate, MedChemExpress

Product Code. 16491996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491996 500 mg 500mg
16468076 100 mg 100mg
16431366 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491996 Supplier Medchem Express Supplier No. HYB0076
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bisoprolol hemifumarate is a potent, selective and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. Bisoprolol hemifumarate has the potential for hypertension, coronary artery disease and stable ventricular dysfunction research.

Bisoprolol hemifumarate is a potent, selective and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. Bisoprolol hemifumarate has the potential for hypertension, coronary artery disease and stable ventricular dysfunction research.

Specifications

Chemical Name or Material Bisoprolol hemifumarate
CAS 104344-23-2
Color White
Purity Grade Notes Research
Molecular Formula C18H31NO4.1/2C4H4O4
Quantity 500 mg
Solubility Information DMSO : ≥ 50 mg/mL (65.19 mM) ∣H2O : 20 mg/mL (26.08 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=C(COCCOC(C)C)C=C1.OC(/C=C/C(O)=O)=O.[1/2]
Molecular Weight (g/mol) 766.96
Formula Weight 766.96
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.