missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol EP Impurity C, Mikromol™

Product Code. 17960826
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
17960826 25 mg 25mg
17940386 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 17960826

Brand: Mikromol MM0460.150025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bisoprolol EP Impurity C
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (RS)-1-[4-[4-(2-Hydroxy-3-isopropylaminopropoxy)benzyl]phenoxy]-3-isopropylaminopropan-2-ol
CAS 1225195-70-9
Analyte or Component Names (RS)-1-[4-[4-(2-Hydroxy-3-isopropylaminopropoxy)benzyl]phenoxy]-3-isopropylaminopropan-2-ol
Shipping Condition Room Temperature
Molecular Formula C25 H38 N2 O4
Certifications/Compliance ISO 17025
Synonym Bisoprolol Fumarate Imp. C (EP), Bisoprolol Imp. C (EP), (RS)-1-[4-[4-(2-Hydroxy-3-isopropylaminopropoxy)benzyl]phenoxy]-3-isopropylaminopropan-2-ol, Bisoprolol Fumarate Impurity C, Bisoprolol Impurity C
InChI Formula InChI=1S/C19H20F3NO2.ClH/c1-14(13-16-9-5-6-10-17(16)19(20,21)22)23-11-12-25-18(24)15-7-3-2-4-8-15;/h2-10,14,23H,11-13H2,1H3;1H
SMILES CC(C)NCC(O)COc1ccc(Cc2ccc(OCC(O)CNC(C)C)cc2)cc1
IUPAC Name 1-[4-[[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]methyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molecular Weight (g/mol) 430.58
Quantity 25 mg
Formula Weight 430.2832 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.