missing translation for 'onlineSavingsMsg'
Learn More

Bisoctrizole, TRC

Product Code. 17937844
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17937844 25 g 25g
30134344 50 g 50g
30134884 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17937844 Supplier Toronto Research Chemicals Supplier No. TRCB50887025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103597-45-1
Molecular Formula C41 H50 N6 O2
Molecular Weight (g/mol) 658.87
Synonym 2,2'-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol], 2,2'-Methylenebis(4-tert-octyl-6-benzotriazol-2-ylphenol), 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-(2H-benzotriazol-2-yl)phenol], 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-benzotriazol-2-ylphenol], 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-benzotriazolylphenol], 2,2'-Methylenebis[6-(2-benzotriazolyl)-4-(1,1,3,3-tetramethylbutyl)phenol], 2,2'-Methylenebis[6-(2H-benzotriazol-2-yl)-4-tert-octylphenol], 2,2'-Methylenebis[6-(2H-benzotriazole-2-yl)-4-tert-octylphenol], ADK Stab LA 31, ADK Stab LA 31R, ADK Stab LA 31RG, ADK-ARKLS LA 31, Bis[2-hydroxy-5-tert-octyl-3-(benzotriazol-2-yl)phenyl]methane, Bisoctrizole, Bisoctyltriazole, Chiguard 5431, Eversorb 78, FAT 75'634, JF 832, Kemisorb 279, LA 31, LA 31RG, MBBT, Mark LA 31, Methylenebis(2-hydroxy-3-(benzotriazol-2-yl)-5-tert-octylphenyl), Mixxim ADK LA 31, Mixxim BB 100, Tinosorb M, Tinuvin 360, UV 360, ZINC 11677911
IUPAC Name 2-(benzotriazol-2-yl)-6-[[3-(benzotriazol-2-yl)-2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]methyl]-4-(2,4,4-trimethylpentan-2-yl)phenol
SMILES CC(C)(C)CC(C)(C)c1cc(Cc2cc(cc(c2O)n3nc4ccccc4n3)C(C)(C)CC(C)(C)C)c(O)c(c1)n5nc6ccccc6n5

Specifications

Quantity 25 g
InChI Formula InChI=1S/C41H50N6O2/c1-38(2,3)24-40(7,8)28-20-26(36(48)34(22-28)46-42-30-15-11-12-16-31(30)43-46)19-27-21-29(41(9,10)25-39(4,5)6)23-35(37(27)49)47-44-32-17-13-14-18-33(32)45-47/h11-18,20-23,48-49H,19,24-25H2,1-10H3
Formula Weight 658.3995
Chemical Name or Material Bisoctrizole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.