missing translation for 'onlineSavingsMsg'
Learn More

Bisoctrizole, Mikromol™

Product Code. 17999874
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17999874

Brand: Mikromol MM3255.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bisoctrizole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisoctrizole
CAS 103597-45-1
Analyte or Component Names Bisoctrizole
Shipping Condition Room Temperature
Molecular Formula C41 H50 N6 O2
Certifications/Compliance ISO 17034
Synonym 2,2'-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol, 2,2'-Methylenebis(4-tert-octyl-6-benzotriazol-2-ylphenol), 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-(2H-benzotriazol-2-yl)phenol], 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-benzotriazol-2-ylphenol], 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-benzotriazolylphenol], 2,2'-Methylenebis[6-(2-benzotriazolyl)-4-(1,1,3,3-tetramethylbutyl)phenol], 2,2'-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol], 2,2'-Methylenebis[6-(2H-benzotriazol-2-yl)-4-tert-octylphenol], ADK Stab LA 31, ADK Stab LA 31R, ADK Stab LA 31RG, ADK-ARKLS LA 31, Adekastab LA 31, Bis[2-hydroxy-5-tert-octyl-3-(benzotriazol-2-yl)phenyl]methane, Bisoctrizole, Bisoctyltriazole, Chiguard 5431, Eversorb 78, FAT 75'634, JF 832, LA 31, LA 31RG, MBBT, Mark LA 31, Methylenebis(2-hydroxy-3-(benzotriazol-2-yl)-5-tert-octylphenyl), Mixxim ADK LA 31, Mixxim BB 100, Tinosorb M, Tinuvin 360, UV 360, ZINC 11677911
InChI Formula InChI=1S/C41H50N6O2/c1-38(2,3)24-40(7,8)28-20-26(36(48)34(22-28)46-42-30-15-11-12-16-31(30)43-46)19-27-21-29(41(9,10)25-39(4,5)6)23-35(37(27)49)47-44-32-17-13-14-18-33(32)45-47/h11-18,20-23,48-49H,19,24-25H2,1-10H3
SMILES CC(C)(C)CC(C)(C)c1cc(Cc2cc(cc(c2O)n3nc4ccccc4n3)C(C)(C)CC(C)(C)C)c(O)c(c1)n5nc6ccccc6n5
IUPAC Name 2-(benzotriazol-2-yl)-6-[[3-(benzotriazol-2-yl)-2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]methyl]-4-(2,4,4-trimethylpentan-2-yl)phenol
Molecular Weight (g/mol) 658.87
Quantity 250 mg
Formula Weight 658.3995 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.