missing translation for 'onlineSavingsMsg'
Learn More

Bismuth vanadium oxide, 99.9% (metals basis)

Product Code. 11333618
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333618 25 g 25g
11343618 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333618 Supplier Thermo Scientific Alfa Aesar Supplier No. 039220.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14059-33-7 | BiO4V | 323.918 g/mol

Bismuth vanadium oxide is used as a pigment due to its hiding power. It also finds use in paints, coatings, plastic and rubber products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14059-33-7
Molecular Formula BiO4V
Molecular Weight (g/mol) 323.918
MDL Number MFCD00064836
InChI Key YNOWMDYNXXVJMC-UHFFFAOYSA-N
Synonym Bismuth vanadate
PubChem CID 20243764
IUPAC Name bismuth;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Bi+3]

Specifications

Color Red to Yellow
Physical Form Powder
Quantity 25 g
UN Number UN3285
Solubility Information Soluble in acids; Insoluble in water.
Formula Weight 323
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Bismuth vanadium oxide
compliance-icons
Product Identifier
  • Bismuth vanadium oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.