missing translation for 'onlineSavingsMsg'
Learn More

Bismuth tungsten oxide, 99.9% (metals basis)

Product Code. 11354817
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354817 25 g 25g
11364817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354817 Supplier Thermo Scientific Alfa Aesar Supplier No. 041854.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13595-87-4 | Bi2O12W3 | 1161.469 g/mol

Acts as a visible-light driven photocatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13595-87-4
Molecular Formula Bi2O12W3
Molecular Weight (g/mol) 1161.469
MDL Number MFCD00046190
InChI Key DWNBMQNJXHTBPG-UHFFFAOYSA-N
Synonym bismuth tungstate, bismuth tungsten oxide, 2bi.3wo4, dibismuth 3+ ion tritungstate, dibismuth;dioxido dioxo tungsten, dibismuth 3+ tritungstate, bismuth tungsten oxide, powder,-200 mesh trace metals basis
PubChem CID 22234745
IUPAC Name dibismuth;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[O-][W](=O)(=O)[O-].[O-][W](=O)(=O)[O-].[Bi+3].[Bi+3]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 1161.48
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Bismuth tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.