missing translation for 'onlineSavingsMsg'
Learn More

Bismuth titanium isopropoxide, 5% w/v in isopropanol, Thermo Scientific™

Product Code. 11380048
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380048 100 mL 100mL
11390048 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380048 Supplier Thermo Scientific Alfa Aesar Supplier No. 042839.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Preparation of thin film mixed oxides of Ti and Bi

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338391-61-0
Molecular Formula C21H49BiO7Ti
MDL Number MFCD01865878
Synonym bismuthane titanium, bismuth-titanium 1/1, bismuth, compd. with titanium 1:1, bismuth titanium isopropoxide w/v in isopropanol, bismuth titanium isopropoxide w/v in isopropanol, packaged under argon in resealable chemseal bottles

Specifications

Color Yellow
Linear Formula BiTi[OCH(CH3)2]7
Quantity 100 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 670.48
Physical Form Liquid
Chemical Name or Material Bismuth titanium isopropoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.