Learn More
Bismuth subsalicylate, 96%, Thermo Scientific Chemicals
£85.20 - £272.00
Chemical Identifiers
CAS | 14882-18-9 |
---|---|
Molecular Formula | C7H6BiO4 |
Molecular Weight (g/mol) | 363.101 |
MDL Number | MFCD00085368 |
InChI Key | QBWLKDFBINPHFT-UHFFFAOYSA-L |
Synonym | bismuth subsalicylate, bismuth oxysalicylate, wismutsubsalicylat, bismuth oxide salicylate, stabisol, vismut, bismuthi subsalicylas, spiromak forte, pepto-bismol, bismutum subsalicylicum |
PubChem CID | 16682734 |
IUPAC Name | 1,3,2$l^{2}-benzodioxabismin-4-one;hydrate |
SMILES | C1=CC=C2C(=C1)C(=O)O[Bi]O2.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11383978
|
Thermo Scientific Alfa Aesar
040395.30 |
250 g |
£85.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393978
|
Thermo Scientific Alfa Aesar
040395.A1 |
1 kg |
£272.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
To impart pearly surface to cellulose-base, polystyrene, and phenolformaldehyde resins. Typical tap bulk density 0.43g/cm^3Bismuth subsalicylate is used to treat temporary discomforts of the stomach and gastrointestinal tract, such as diarrhea, indigestion, heartburn and nausea. It also acts as an antacid. It is practiced to decrease the number of bowel movements and to make the stool firmer. It is also used to reduce inflammation within the intestine. It is further used in growth media for selective isolation of microorganisms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14882-18-9 | |
363.101 | |
QBWLKDFBINPHFT-UHFFFAOYSA-L | |
16682734 | |
C1=CC=C2C(=C1)C(=O)O[Bi]O2.O |
C7H6BiO4 | |
MFCD00085368 | |
bismuth subsalicylate, bismuth oxysalicylate, wismutsubsalicylat, bismuth oxide salicylate, stabisol, vismut, bismuthi subsalicylas, spiromak forte, pepto-bismol, bismutum subsalicylicum | |
1,3,2$l^{2}-benzodioxabismin-4-one;hydrate |
Specifications
14882-18-9 | |
Powder | |
C7H6BiO4 | |
bismuth subsalicylate, bismuth oxysalicylate, wismutsubsalicylat, bismuth oxide salicylate, stabisol, vismut, bismuthi subsalicylas, spiromak forte, pepto-bismol, bismutum subsalicylicum | |
QBWLKDFBINPHFT-UHFFFAOYSA-L | |
1,3,2$l^{2}-benzodioxabismin-4-one;hydrate | |
16682734 | |
Odorless | |
Bismuth subsalicylate |
White | |
96% | |
MFCD00085368 | |
Practically insoluble in water,alcoholSoluble in acid and alkali. Insoluble in cold water,ethanol,diethyl ether and n-octanol. | |
C1=CC=C2C(=C1)C(=O)O[Bi]O2.O | |
363.101 | |
362.09 | |
White powder |
Safety and Handling
H315-H319-H335
missing translation for 'einecsNumber' : 238-953-1
missing translation for 'rtecsNumber' : EB2985000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Bismuth subsalicylate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only