missing translation for 'onlineSavingsMsg'
Learn More

Bismuth powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11326867
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray powder
Assay Percent Range 99.999% (Metals basis)
Quantity 25 g
Mesh Size -200 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Bismuth
compliance-icons
Product Identifier
  • Bismuth powder
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'