missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth, plasma standard solution, Specpure™ Bi 10μg/mL, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Concentration | Bi in 2% HNO3 |
Quantity | 100 mL |
MDL Number | MFCD00134033 |
UN Number | UN3264 |
Grade | Specpure™ |
Physical Form | Liquid |
Chemical Name or Material | Bismuth, plasma standard solution, Bi 10μg/mL |
Sécurité et traitement
Identifiant produit
- Bismuth, plasma standard solution, Specpure, Bi 10µg/ml
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'