missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Nitrate Pentahydrate, TRC

Product Code. 30337478
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337478 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337478 Supplier Toronto Research Chemicals Supplier No. TRCB4973001G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bismuth Nitrate Pentahydrate
CAS 10035-06-0
Quantity 1 g
Molecular Formula BiH13N3O14
Synonym Bismuth(3+) Salt Nitric Acid Hydrate (3:1:5), Bismuth(3+) Salt Nitric Acid Pentahydrate (3:1:5), Bismuth Trinitrate Pentahydrate, Bismuth(III) Nitrate Pentahydrate, Granions de Bismuth
InChI Formula InChI=1S/Bi.3NO3.5H2O/c;3*2-1(3)4;;;;;/h;;;;5*1H2/q+3;3*-1;;;;;
SMILES O.O.O.O.O.[Bi+3].[O-][N+](=O)[O-].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
IUPAC Name bismuth;trinitrate;pentahydrate
Molecular Weight (g/mol) 488.1
Formula Weight 484.9967
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.