missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Nitrate Pentahydrate, TRC

Product Code. 30330653
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330653 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330653 Supplier Toronto Research Chemicals Supplier No. TRCB49730010G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bismuth Nitrate Pentahydrate
CAS 10035-06-0
Quantity 10 g
Molecular Formula BiH13N3O14
Synonym Bismuth(3+) Salt Nitric Acid Hydrate (3:1:5), Bismuth(3+) Salt Nitric Acid Pentahydrate (3:1:5), Bismuth Trinitrate Pentahydrate, Bismuth(III) Nitrate Pentahydrate, Granions de Bismuth
InChI Formula InChI=1S/Bi.3NO3.5H2O/c;3*2-1(3)4;;;;;/h;;;;5*1H2/q+3;3*-1;;;;;
SMILES O.O.O.O.O.[Bi+3].[O-][N+](=O)[O-].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
IUPAC Name bismuth;trinitrate;pentahydrate
Molecular Weight (g/mol) 488.1
Formula Weight 484.9967
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.