missing translation for 'onlineSavingsMsg'
Learn More

Bismuth needles, 1-5cm (0.4-2in) long, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11331158
Click to view available options
Quantity:
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7440-69-9
Molecular Formula Bi
MDL Number MFCD00134033
Quantity 2 kg
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
SMILES [Bi]
IUPAC Name bismuth
Molecular Weight (g/mol) 208.98
PubChem CID 5359367
ChEBI CHEBI:33301
Percent Purity 99.998%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Bismuth needles, 1 to 5cm (0.4 to 2 in.) long
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'