missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth Lead Tin ingot (Rose's metal), Thermo Scientific Chemicals
87.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
Rose's metal was used as the droplet fluid, because it has a low melting point (e460 K). Rose's metal is used as a sample in computer-controlled diffractometer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 95°C to 96°C |
Avg. Mol. Wt. or Mol. Wt. Range | Bi:Pb:Sn; 50:25:25 wt% |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Form | Melting Alloy |
Assay | (Rose's metal) |
Chemical Name or Material | Bismuth Lead Tin ingot (Rose's metal) |
Safety and Handling
Product Identifier
- Bismuth Lead Tin ingot (Rose's metal)
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'