missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Lead Tin Cadmium ingot (Wood's metal)

Product Code. 11347528
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347528 2 kg 2kg
11357528 100 g 100g
11367528 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347528 Supplier Thermo Scientific Alfa Aesar Supplier No. 033218.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8049-22-7 | Cd12Sn2 | 1586.388 g/mol

Wood's metal is useful as a low-melting solder, low-temperature casting metal, high temperature coupling fluid in heat baths, and as a fire-melted valve element in fire sprinkler systems in buildings. It is also used as a filler when bending thin-walled metal tube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8049-22-7
Molecular Formula Cd12Sn2
Molecular Weight (g/mol) 1586.388
MDL Number MFCD00198175
InChI Key OIBLPQFTAJDRMC-UHFFFAOYSA-N
Synonym lambda∼2∼-stannane-cadmium 1/6
PubChem CID 71434152
IUPAC Name cadmium;tin
SMILES [Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Sn].[Sn]

Specifications

Color Gray
Melting Point 71°C
Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:Sn:Cd; 50:25:12.5:12.5 wt%
Quantity 2 kg
Odor Odorless
Form Ingot
UN Number UN3077
Solubility Information Insoluble in water.
Assay (Wood's metal)
Chemical Name or Material Bismuth Lead Tin Cadmium ingot (Wood's metal)
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Carcinogenicity Category 1B
  • Reproductive toxicity on or via lactation
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H362-May cause harm to breast-fed children.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.