missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) telluride, 99.98% (metals basis)

Product Code. 11395158
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11395158

Brand: Thermo Scientific Alfa Aesar 044077.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1304-82-1 | Bi2Te3

Bismuth(III) telluride and its alloys (antimony or selenium) posses excellent semiconductor activity, which is used in thermoelectric industry for cooling and energy generation. It increases the speed of microchips and acts as topological insulators in spintronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-82-1
Molecular Formula Bi2Te3
MDL Number MFCD00014201

Specifications

Melting Point 573°C
Color Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.98% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 800.76
Odor Odorless
Chemical Name or Material Bismuth(III) telluride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.