missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) sulfate, 99%

Product Code. 15443237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443237 100 g 100g
15433237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443237 Supplier Thermo Scientific Alfa Aesar Supplier No. B22007.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7787-68-0 | Bi2O12S3 | 706.13 g/mol

The esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.

Solubility
Soluble in acid, insoluble in water and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-68-0
Molecular Formula Bi2O12S3
Molecular Weight (g/mol) 706.13
MDL Number MFCD00036140
InChI Key BEQZMQXCOWIHRY-UHFFFAOYSA-H
Synonym bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4
PubChem CID 198144
IUPAC Name dibismuth;trisulfate
SMILES [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 405°C (decomposition)
Quantity 100 g
Density 5.08
Linear Formula Bi2(SO4)3
Merck Index 14,1286
Solubility Information Soluble in acid,insoluble in water and ethanol.
Formula Weight 706.13
Percent Purity 99%
Chemical Name or Material Bismuth(III) sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.