missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(Iii) Subnitrate, Honeywell Fluka™
BIOPHARMA

Product Code. 15680970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
50 kg
Unit Size:
1kg
250g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15680970 1 kg 1kg
15690970 250 g 250g
15600980 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15680970 Supplier Honeywell-Fluka Supplier No. 103341KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of DAB, USP, 71-74% Bi basis (calc. on dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-85-4
MDL Number MFCD00064844

Specifications

Quantity 1 kg
Linear Formula Bi5O(OH)9(NO3)4
UN Number 1477
Formula Weight 1461.99g/mol
Grade puriss.
Concentration or Composition (by Analyte or Components) 71 to 74% Bi basis (calc. on dried substance)
Packaging Plastic Bottle
Chemical Name or Material Bismuth(Iii) Subnitrate, Meets analytical specification of USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.