missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) selenide, 99.995% (metals basis), Thermo Scientific™

Product Code. 15482907
Click to view available options
Quantity:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12068-69-8
MDL Number MFCD00014200
Synonym dibismuth selenium 2-, dibismuth triselenide, dibismuth 3+ ion triselenide, bismuth selenide,, bismuth iii selenide trace metals basis 10g

Specifications

Quantity 1g
UN Number UN3283
Merck Index 14,1278
Solubility Information Insoluble in water.
Formula Weight 654.84
Percent Purity 99.999%
Density 6.82 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) selenide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) selenide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P403-Store in a well-ventilated place.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'