missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) phosphate, 99%

Product Code. 11338158
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338158 50 g 50g
11328158 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338158 Supplier Thermo Scientific Alfa Aesar Supplier No. 039380.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-01-1 | BiH3O4P | 306.97 g/mol

Bismuth(III) phosphate is used in treatment of renal cysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-01-1
Molecular Formula BiH3O4P
Molecular Weight (g/mol) 306.97
MDL Number MFCD00049721
InChI Key SFOQXWSZZPWNCL-UHFFFAOYSA-N
Synonym bismuth phosphate, bismuth iii phosphate, bismugel, phosphoric acid, bismuth 3+ salt 1:1, bismuth phosphate mi, bismuth phosphate who-dd, acmc-20allc, bismuth orthophosphate, bi.po4, bismuth phosphate,
PubChem CID 9839429
SMILES [Bi+3].OP(O)(O)=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 50 g
Merck Index 14,1275
Solubility Information Insoluble in water and alkali; Soluble in hydrochloric acid
IUPAC Name bismuth;phosphate
Formula Weight 303.95
Percent Purity 99%
Odor Odorless
Density 6.32 g/mL
Chemical Name or Material Bismuth(III) phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.