missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxyiodide, 98%, pure, Thermo Scientific™

Product Code. 15259866 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15259866 10 g 10g
15269866 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15259866 Supplier Thermo Scientific Acros Supplier No. 453830100
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7787-63-5
Molecular Formula BiIO
Molecular Weight (g/mol) 351.88
InChI Key FCBARUADRQNVFT-UHFFFAOYSA-N
Synonym bismuth iodide oxide, biio, bismuth iii oxyiodide, bismuth oxyiodide, bismuth oxyiodide, bioi
PubChem CID 6367235
IUPAC Name oxobismuth;hydroiodide
SMILES O=[Bi].I

Specifications

Quantity 10 g
Assay Percent Range 97.5% min
Linear Formula IBi=O
Merck Index 15,127
Solubility Information Solubility in water: insoluble.
Formula Weight 351.88
Percent Purity 98%
Grade Pure
Chemical Name or Material Bismuth(III) oxyiodide
compliance-icons
Product Identifier
  • Bismuth(III) oxyiodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.