Learn More
Bismuth(III) oxide, typically 99.99% (metals basis)
Description
Used as an adhesive and sealant chemical, flame retardant, pigment, propellant and blowing agent. It is also used as a solvent. Other uses are in batteries, electrical and electronic products, explosive materials, ink, toner, and colorant products, paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | ∽1890°C |
| Melting Point | 817°C |
| Physical Form | Powder |
| Quantity | 250 g |
| Assay Percent Range | 99.99% (Metals Basis) |
| Solubility Information | Insoluble in water. Soluble in acids. |
| Formula Weight | 465.96 |
| Density | 8.9 |
| Chemical Name or Material | Bismuth(III) oxide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.