missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 99%

Product Code. 11381579
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381579 1 kg 1kg
11301589 250 g 250g
11391579 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381579 Supplier Thermo Scientific Alfa Aesar Supplier No. 012230.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

It finds use in disinfectants, magnets, glass, rubber, vulcanization, fireproofing papers and polymers, and catalysts. Bismuth trioxide brings about the dragon's eggs effect in fireworks, as a replacement of red lead. Bismuth(III) compounds are attractive reagents and catalysts in organic synthesis because of their low cost and ease of handling. Bismuth trioxide nanoparticles also play an important role in high energy gas generator. The alpha crystalline form of bismuth(III) oxide has p-type electronic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ca 1890°C
Melting Point 817°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99%
Solubility Information Practically insoluble in water. Soluble in acids.
Formula Weight 465.96
Density 8.9 g/cm3
Chemical Name or Material Bismuth(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.