missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 99.9%, (trace metal basis)

Product Code. 10398340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10398340 500 g Plastic bottle 500g
10451441 100 g Plastic bottle 100g
11954161 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10398340 Supplier Thermo Scientific Acros Supplier No. 208075000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
Synonym Bismuth trioxide
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point 1890°C
Melting Point 825.0°C
Color Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9%
Linear Formula Bi2O3
Fieser 01,59
Merck Index 15,1273
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 465.95
Percent Purity 99.9%
Grade Extra Pure
Loss on Drying 0.2% max.
Packaging Plastic bottle
Specific Gravity 8.9
Density 8.9000g/mL
Chemical Name or Material Bismuth(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.