missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) nitrate pentahydrate, 98%

Product Code. 11429293
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429293 500 g 500g
11419293 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429293 Supplier Thermo Scientific Alfa Aesar Supplier No. A11748.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10035-06-0 | BiH10N3O14 | 485.07 g/mol

Bismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

Solubility
Soluble in dilute nitric acid solution, dilute acetic acid, glycerol and acetone. Insoluble in alcohol, ethyl acetateSoluble in acetone, acetic acid and glycerol. Insoluble in ethanol and ethyl acetate.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong acids, strong reducing agents, strong oxidizing agents, powdered metals and organic materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-06-0
Molecular Formula BiH10N3O14
Molecular Weight (g/mol) 485.07
MDL Number MFCD00149157
InChI Key FBXVOTBTGXARNA-UHFFFAOYSA-N
Synonym bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
PubChem CID 160911
IUPAC Name bismuth;trinitrate;pentahydrate
SMILES O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 30°C (decomposition)
Quantity 500 g
UN Number UN1477
Merck Index 14,1271
Solubility Information Soluble in dilute nitric acid solution,dilute acetic acid,glycerol and acetone. Insoluble in alcohol,ethyl acetateSoluble in acetone,acetic acid and glycerol. Insoluble in ethanol and ethyl acetate.
Formula Weight 485.07 (394.99 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.83
Chemical Name or Material Bismuth (III) nitrate pentahydrate
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H372-Causes damage to organs.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.