missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) nitrate pentahydrate, 98%, Thermo Scientific Chemicals

Catalog Number 11419293
Click to view available options
Quantity:
100 g
500 g
10035-06-0
BiH10N3O14
485.07
MFCD00149157
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
160911
bismuth;trinitrate;pentahydrate
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
10035-06-0
BiH10N3O14
485.07
MFCD00149157
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
160911
bismuth;trinitrate;pentahydrate
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Bismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

Solubility
Soluble in dilute nitric acid solution, dilute acetic acid, glycerol and acetone. Insoluble in alcohol, ethyl acetateSoluble in acetone, acetic acid and glycerol. Insoluble in ethanol and ethyl acetate.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong acids, strong reducing agents, strong oxidizing agents, powdered metals and organic materials.
TRUSTED_SUSTAINABILITY
Melting Point 30°C (decomposition)
Quantity 100 g
UN Number UN1477
Merck Index 14,1271
Solubility Information Soluble in dilute nitric acid solution,dilute acetic acid,glycerol and acetone. Insoluble in alcohol,ethyl acetateSoluble in acetone,acetic acid and glycerol. Insoluble in ethanol and ethyl acetate.
Formula Weight 485.07 (394.99 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.83
Chemical Name or Material Bismuth (III) nitrate pentahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'