missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11370869
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11370869 100 g 100g
11380869 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11370869 Supplier Thermo Scientific Alfa Aesar Supplier No. 010657.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10361-44-1 | BiN3O9 | 394.99 g/mol

It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-44-1
Molecular Formula BiN3O9
Molecular Weight (g/mol) 394.99
MDL Number MFCD00003460
InChI Key PPNKDDZCLDMRHS-UHFFFAOYSA-N
Synonym bismuth nitrate, bismuth trinitrate, ccris 7110, nitric acid, bismuth 3+ salt, tris nitrooxy bismuthine, bismuth 3+ trinitrate, bi.3no3, bismuth iii nitrate, bismuth iii nitrate hydrate, nitric acid, bismuth 3+ salt 3:1
PubChem CID 107711
IUPAC Name bismuth;trinitrate
SMILES [Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 30°C (decomposition)
Color Colorless to White
Physical Form Crystalline
Quantity 100 g
UN Number UN1477
Merck Index 14,1271
Solubility Information Slightly soluble in water; soluble in acetone,acetic acid,and glycerol. Insoluble in ethyl acetate and ethanol.
Formula Weight 394.99 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Bismuth(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.