missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) fluoride, anhydrous, 99%

Product Code. 11382168
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382168 500 g 500g
11372168 25 g 25g
11392168 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382168 Supplier Thermo Scientific Alfa Aesar Supplier No. 013075.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufacturing of bismuth pentafluoride | CAS: 7787-61-3 | BiF3 | 265.976 g/mol

It is used in the manufacturing of bismuth pentafluoride. Bismuth(III) fluoride is an attractive reagent and catalyst in organic synthesis owing to its low cost, and ease of handling. It is also used in polarography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-61-3
Molecular Formula BiF3
Molecular Weight (g/mol) 265.976
MDL Number MFCD00010893
InChI Key BRCWHGIUHLWZBK-UHFFFAOYSA-K
Synonym bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
PubChem CID 82233
IUPAC Name trifluorobismuthane
SMILES F[Bi](F)F

Specifications

Melting Point 727°C
Physical Form Powder
Quantity 500 g
UN Number UN3260
Merck Index 14,1265
Solubility Information Insoluble in water; Soluble in HF.
Formula Weight 265.98
Percent Purity 99%
Odor Odorless
Refractive Index 1.74
Sensitivity Hygroscopic
Chemical Name or Material Bismuth(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.