missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bismuth(III) fluoride, anhydrous, 99%

Manufacturing of bismuth pentafluoride

Brand:  Alfa Aesar™ 013075.22

Code : NEW

Additional Details : CAS Number : 7787-61-3 Weight : 0.10000kg Transport : UN number : 3260 Chem class : 1a Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11392168

  • £78.90 / 100g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

Bismuth(III) fluoride
Anhydrous
MFCD00010893
14,1265
7787-61-3
265.98
UN3260

Specifications

Bismuth(III) fluoride
7787-61-3
99%
MFCD00010893
14,1265
Odorless
Hygroscopic
100g
Anhydrous
Powder
265.98
UN3260
Practically insoluble in water; Soluble in HF
1.74
727°C
Videos
SDS