missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) chloride, ultra dry, 99.997+% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Bismuth chloride is used as a catalyst in organic synthesis. In particular, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. The addition of other metal iodides increases its catalytic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | ∼430°C (sublimation) |
Melting Point | 230°C to 232°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN3260 |
Merck Index | 14,1262 |
Solubility Information | It is soluble in methanol, diethyl ether, acetone. |
Formula Weight | 315.34 |
Percent Purity | ≥99.997% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'