missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) Chloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Bismuth(III) Chloride |
| CAS | 7787-60-2 |
| Molecular Formula | BiCl3 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/Bi.3ClH/h;3*1H/q+3;;;/p-3 |
| SMILES | [Cl-].[Cl-].[Cl-].[Bi+3] |
| IUPAC Name | bismuth;trichloride |
| Molecular Weight (g/mol) | 315.34 |
| Formula Weight | 313.8869 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction