missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride oxide, 98%, Alfa Aesar™

Product Code. 15472537
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472537 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15472537 Supplier Thermo Scientific Alfa Aesar Supplier No. 045899.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7787-59-9
Physical Form Chunks
Quantity 50 g
Molecular Formula BiClHO
Linear Formula BiOCl
MDL Number MFCD00010895
Synonym bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid
InChI Key BWOROQSFKKODDR-UHFFFAOYSA-N
SMILES O=[Bi].Cl
IUPAC Name oxobismuth;hydrochloride
Molecular Weight (g/mol) 261.437
PubChem CID 6328152
Formula Weight 260.43
Percent Purity 98%
Packaging Plastic bottle
Chemical Name or Material Bismuth(III) chloride oxide
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) chloride oxide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.