missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
70.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
It is used as a catalyst in organic chemistry. It is a good source of bismuth and is used to make other bismuth compounds. Also used as a catalyst and to make other bismuth salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | ∼430°C (sublimation) |
Melting Point | 230°C to 232°C |
Color | White |
Physical Form | Crystalline |
Quantity | 5 g |
UN Number | UN3260 |
Merck Index | 14,1262 |
Solubility Information | Soluble in water. |
Formula Weight | 315.34 |
Percent Purity | 99.999% |
Show More |
Safety and Handling
Product Identifier
- Bismuth(III) chloride
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'