missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) bromide, anhydrous, 99.999% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 047175.06

Quantity 5g

 View more versions of this product

Product Code. 15427445

  • £289.00 / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7787-58-8
448.692
TXKAQZRUJUNDHI-UHFFFAOYSA-K
82232
Br[Bi](Br)Br
BiBr3
MFCD00014195
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
tribromobismuthane

Specifications

7787-58-8
BiBr3
UN3260
Decomposes in water.
Br[Bi](Br)Br
448.692
448.71
Acidic
5.72 g/mL
Bismuth(III) bromide, Anhydrous
5g
MFCD00014195
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
TXKAQZRUJUNDHI-UHFFFAOYSA-K
tribromobismuthane
82232
99.999%
Hygroscopic
(metals basis)
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.