missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) acetate, 99.999% (metals basis)

Product Code. 11311918
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311918 10 g 10g
11301918 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311918 Supplier Thermo Scientific Alfa Aesar Supplier No. 011846.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 22306-37-2 | C6H9BiO6 | 386.112 g/mol

It is used in biology, printing, in photographic films, textiles, and also in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22306-37-2
Molecular Formula C6H9BiO6
Molecular Weight (g/mol) 386.112
MDL Number MFCD00015629
InChI Key WKLWZEWIYUTZNJ-UHFFFAOYSA-K
Synonym bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g
PubChem CID 31132
IUPAC Name bismuth;triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3]

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystals
Quantity 10 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 386.12
Sensitivity Hygroscopic
Chemical Name or Material Bismuth(III) acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.