missing translation for 'onlineSavingsMsg'
Learn More

Bismuth hydroxide, tech.

Product Code. 11343988
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343988 250 g 250g
11333988 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343988 Supplier Thermo Scientific Alfa Aesar Supplier No. 040397.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrolysis of ribonucleic acid, absorbent | CAS: 10361-43-0 | BiH3O3 | 260.00 g/mol

It is used for the hydrolysis of ribonucleic acid and as absorbent for rutin and quercetin. It is a component used in milk of bismuth is used in gastrointestinal disorders as a protective agent. It is used for the removal of nitrate and other anions from water by yellow bismuth hydroxide. A bismuth oxide nanoplate is useful for carbon dioxide gas sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-43-0
Molecular Formula BiH3O3
Molecular Weight (g/mol) 260.00
MDL Number MFCD00046188
InChI Key TZSXPYWRDWEXHG-UHFFFAOYSA-K
IUPAC Name bismuth(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Bi+3]

Specifications

Physical Form Amorphous Powder
Quantity 250 g
Solubility Information Insoluble in water; Soluble in acids.
Formula Weight 260
Grade Technical
Odor Odorless
Chemical Name or Material Bismuth hydroxide
compliance-icons
Product Identifier
  • Bismuth hydroxide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.