missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is used pharmaceutically in wound dressing, ointment component and as an antiacid. In cosmetics it is used as a cosmetic white pigment. Used in Ceramic glazes and enamel fluxes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 260°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN1477 |
Merck Index | 14,1284 |
Solubility Information | Practically insoluble in water and alcohol; Soluble in dilute HNO3 and HCl |
Formula Weight | 1461.98 |
Concentration or Composition (by Analyte or Components) | Bi2O3 79% min. |
Odor | Odorless |
Show More |
Safety and Handling
Product Identifier
- Bismuth hydroxide nitrate oxide
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'