missing translation for 'onlineSavingsMsg'
Learn More

Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min

Product Code. 11368448
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368448 100 g 100g
11358448 25 g 25g
11348448 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368448 Supplier Thermo Scientific Alfa Aesar Supplier No. 040476.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ceramic glazes and enamel fluxes | CAS: 1304-85-4

It is used pharmaceutically in wound dressing, ointment component and as an antiacid. In cosmetics it is used as a cosmetic white pigment. Used in Ceramic glazes and enamel fluxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-85-4
MDL Number MFCD00064844
Synonym Bismuth subnitrate

Specifications

Melting Point 260°C (decomposition)
Color White
Physical Form Powder
Quantity 100 g
UN Number UN1477
Merck Index 14,1284
Solubility Information Practically insoluble in water and alcohol; Soluble in dilute HNO3 and HCl
Formula Weight 1461.98
Concentration or Composition (by Analyte or Components) Bi2O3 79% min.
Odor Odorless
Sensitivity Hygroscopic
Density 6.04 g/mL
Chemical Name or Material Bismuth hydroxide nitrate oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bismuth hydroxide nitrate oxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: Bismuth hydroxide nitrate oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.