missing translation for 'onlineSavingsMsg'
Learn More

Bismuth foil, 2.0mm (0.08 in.) thick, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11450411
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450411 50 x 50 mm Each
11328778 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11450411 Supplier Thermo Scientific Alfa Aesar Supplier No. 041637.FI
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Quantity 50 x 50 mm
Odor Odorless
Weight ≈12.3g/25x25mm
Form Foil
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Bismuth foil, 2.0mm (0.08 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.