missing translation for 'onlineSavingsMsg'
Learn More

Bismuth citrate, 94%

Product Code. 11353988
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11353988

Brand: Thermo Scientific Alfa Aesar 040398.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 813-93-4 | C6H5BiO7 | 398.079 g/mol

Bismuth citrate is used as a progressive hair dye in cosmetics. It is used as antimicrobial agents for syphills, colitis, wound infection, quartan malaria, dyspepsia and peptic ulcers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-93-4
Molecular Formula C6H5BiO7
Molecular Weight (g/mol) 398.079
MDL Number MFCD00050817
InChI Key ANERHPOLUMFRDC-UHFFFAOYSA-K
Synonym bismuth citrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, bismuth 3+ salt 1:1, bismuth citrate usp:usp, bismuth iii citrate, bismuth citrate usp, bismuth 3+ ion citrate, bismuth, citrato 3-, bismuth iii citrate 250g, bismuth 2-hydroxypropane-1,2,3-tricarboxylate
PubChem CID 13135
IUPAC Name bismuth;2-hydroxypropane-1,2,3-tricarboxylate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.[Bi+3]

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Solubility Information Insoluble in water,alcohol,ether; Soluble in ammonia solution and in solution of alkali citrates; Typical tap density 0;50g/cm3;Soluble in ammonia,alkali citrates; Insoluble in water,alcohol and ether.
Formula Weight 398.08
Percent Purity 94%
Odor Odorless
Sensitivity Light sensitive
Density 3.5 g/mL
Chemical Name or Material Bismuth citrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.