missing translation for 'onlineSavingsMsg'
Learn More

Bismuth aluminum oxide hydrate, 95.5%, Thermo Scientific™

Product Code. 11343978
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343978 50 g 50g
11323978 250 g 250g
11333978 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343978 Supplier Thermo Scientific Alfa Aesar Supplier No. 040393.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00798526
Synonym Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water; Soluble in nitric acid; Typical tap density 0;69g/cm3
Formula Weight 771.84 (Anhydrous)
Percent Purity 95.5%
Chemical Name or Material Bismuth aluminum oxide hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.