missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth 2-ethylhexanoate, 99.99+%, 70% w/v in xylene, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 5 g |
UN Number | UN1307 |
Formula Weight | 638.61 |
Percent Purity | ≥99.99% |
Physical Form | Liquid |
Chemical Name or Material | Bismuth 2-ethylhexanoate |
Safety and Handling
Product Identifier
- Bismuth 2-ethylhexanoate 70% w/v in xylene
- Warning
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 2
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H361-Suspected of damaging fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P262-Do not get in eyes, on skin, or on clothing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'