missing translation for 'onlineSavingsMsg'
Learn More

Bismuth 2-ethylhexanoate, 99.99+%, 70% w/v in xylene, Thermo Scientific™

Product Code. 11334268
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334268 100 g 100g
11344268 5 g 5g
11324268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334268 Supplier Thermo Scientific Alfa Aesar Supplier No. 041521.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67874-71-9
Molecular Formula C24H48BiO6
Molecular Weight (g/mol) 641.622
MDL Number MFCD00043803
InChI Key MWXWNYHYPMRCHN-UHFFFAOYSA-N
Synonym 2-Ethylhexanoic acid bismuth salt
PubChem CID 131857451
IUPAC Name bismuth;2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]

Specifications

Quantity 100 g
UN Number UN1307
Formula Weight 638.61
Percent Purity ≥99.99%
Physical Form Liquid
Chemical Name or Material Bismuth 2-ethylhexanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.