missing translation for 'onlineSavingsMsg'
Learn More

Bismarck Brown Y MP Biomedicals

Product Code. 11427721 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427721 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11427721 Supplier MP Biomedicals Supplier No. 0215047380
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plasma stain used for staining mucine, cartilage, and goblet cells.

Specifications

CAS 10114-58-6
Assay Percent Range ∽ 50%
Molecular Formula C18H20Cl2N8
Synonym 4,4-[1,3-PhenyleneBis(Azo)Bis]-1,3-Benzediamine Dihydrochloride, Basic Brown 1
Color Orange
Quantity 100 g
InChI Key MCZVRBLCRZWFJH-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl
IUPAC Name 4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride
Molecular Weight (g/mol) 419.314
PubChem CID 82360
ChEBI CHEBI:53615
Formula Weight 419.3
Grade Not Available
Chemical Name or Material Bismarck Brown Y, Plasma stain for mucin, cartilage and goblet cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.