missing translation for 'onlineSavingsMsg'
Learn More

Bisindolylmaleimide 1, Thermo Scientific Chemicals

Product Code. 15475649
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475649 5 mg 5mg
15485649 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475649 Supplier Thermo Scientific Alfa Aesar Supplier No. J63401.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Very potent and selective inhibitor of protein kinase C

Bisindolylmaleimide is used as a highly selective inhibitor of all PKC isoforms and slight inhibitor of GSK-3, used to selectively probe for PKC-mediated pathways for transduction of hormone, cytokine, and growth factor signals, very potent and selective inhibitor of protein kinase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bisindolylmaleimide is used as a highly selective inhibitor of all PKC isoforms and slight inhibitor of GSK-3, used to selectively probe for PKC-mediated pathways for transduction of hormone, cytokine, and growth factor signals, very potent and selective inhibitor of protein kinase C.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133052-90-1
Molecular Formula C25H24N4O2
Molecular Weight (g/mol) 412.493
MDL Number MFCD00236428
InChI Key QMGUOJYZJKLOLH-UHFFFAOYSA-N
Synonym bisindolylmaleimide i, unii-l79h6n0v6c, rbt205 inhibitor, gö 6850, bisindolylmaleimide i gf 109203x, bim-1, chembl7463, 2-1-3-dimethylaminopropyl indol-3-yl-3-indol-3-yl maleimide, 3-1-3-dimethylamino propyl-1h-indol-3-yl-4-1h-indol-3-yl-1h-pyrrole-2,5-dione
PubChem CID 2396
ChEBI CHEBI:41072
IUPAC Name 3-[1-[3-(dimethylamino)propyl]indol-3-yl]-4-(1H-indol-3-yl)pyrrole-2,5-dione
SMILES CN(C)CCCN1C=C(C2=CC=CC=C21)C3=C(C(=O)NC3=O)C4=CNC5=CC=CC=C54

Specifications

Color Orange
Quantity 5 mg
Solubility Information Insoluble in water.
Formula Weight 412.48
Physical Form Crystalline Powder
Chemical Name or Material Bisindolylmaleimide 1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.