missing translation for 'onlineSavingsMsg'
Learn More

Bisantrene, MedChemExpress

Product Code. 16413208
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16413208

Brand: Medchem Express HY100875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bisantrene is a highly effective antitumor drug, targets eukaryotic type II topoisomerases.

Bisantrene is a highly effective antitumor drug, targets eukaryotic type II topoisomerases.

Specifications

Chemical Name or Material Bisantrene
Physical Form Solid
CAS 78186-34-2
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H22N8
For Use With (Application) Cancer-programmed cell death
Synonym CL216942
Solubility Information DMSO : 1 mg/mL (2.51 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 0.1 mg/mL (0.25 mM; ultrasonic and warming and heat to 50°C)
SMILES C12=CC=CC=C1C(/C=N/NC3=NCCN3)=C4C(C=CC=C4)=C2/C=N/NC5=NCCN5
Molecular Weight (g/mol) 398.46
Formula Weight 398.46
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.