missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIS-TRIS, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | BIS-TRIS |
| CAS | 6976-37-0 |
| Molecular Formula | C8H19NO5 |
| Quantity | 5 g |
| Synonym | Bis-(2-hydroxyethyl)aminotris(hydroxymethyl)methane |
| SMILES | OCCN(CCO)C(CO)(CO)CO |
| Molecular Weight (g/mol) | 209.24 |
| Formula Weight | 209.1263 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction