missing translation for 'onlineSavingsMsg'
Learn More

BIS-TRIS, CellPURE™, Fisher BioReagents

Product Code. 10440060
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10440060 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10440060 Supplier Fisher BioReagents Supplier No. BP2941100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6976-37-0
Melting Point 103°C
Color White
pH 8.8 to 9.6
Physical Form Powder or Solid
Quantity 100 g
Assay Percent Range ≥98.0 %
Infrared Spectrum Conforms to reference
Molecular Formula C8H19NO5
Packaging Poly Bottle
Vapor Pressure Negligible
Synonym 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1, 3-propanediol
Solubility Information Soluble in water
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
SMILES C(CO)N(CCO)C(CO)(CO)CO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 209.242
PubChem CID 81462
ChEBI CHEBI:41250
Formula Weight 209.24
Percent Purity ≥98.0%
Grade CellPURE
Chemical Name or Material BIS-TRIS, Cell culture tested
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.