missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals BIS-TRIS, 0.2M buffer soln., pH 6.0
£63.10 - £98.70
Chemical Identifiers
CAS | 6976-37-0 |
---|---|
Molecular Formula | C8H19NO5 |
Molecular Weight (g/mol) | 209.242 |
MDL Number | MFCD00002853 |
InChI Key | OWMVSZAMULFTJU-UHFFFAOYSA-N |
PubChem CID | 81462 |
ChEBI | CHEBI:41250 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(CO)N(CCO)C(CO)(CO)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435009
|
Thermo Scientific Chemicals
J62686.AE |
100 mL |
£63.10
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445009
|
Thermo Scientific Chemicals
J62686.AK |
250 mL |
£98.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6976-37-0 | |
209.242 | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
CHEBI:41250 | |
C(CO)N(CCO)C(CO)(CO)CO |
C8H19NO5 | |
MFCD00002853 | |
81462 | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
Specifications
6976-37-0 | |
0.2 M | |
Liquid | |
Odorless | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
81462 | |
BIS-TRIS |
Colorless | |
6.0 | |
C8H19NO5 | |
MFCD00002853 | |
C(CO)N(CCO)C(CO)(CO)CO | |
209.242 | |
CHEBI:41250 |
Safety and Handling
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only